siRNA Details
| Gene Symbol: Glb1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192018.1 | NP_001178947.1 | 20 | 1941 | ||
| XM_006242738.2 | XP_006242800.1 | 19 | 1979 | |||
| XM_008766062.1 | XP_008764284.1 | 21 | 3071 | |||
| XM_008766063.1 | XP_008764285.1 | 20 | 1955 | |||
| XM_008766064.1 | XP_008764286.1 | 19 | 2051 | |||

Glb1l2
Genomic Map