siRNA Details
| Gene Symbol: Rbpms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173426.1 | NP_001166897.1 | 5, 6 | 412 | ||
| XM_006243274.2 | XP_006243336.1 | 5, 6 | 2098 | |||
| XM_008766329.1 | XP_008764551.1 | 4, 5 | 521 | |||
| GenBank mRNA | BC169038.1 | - | 5, 6 | 412 | 9090232 | |

Rbpms2
Genomic Map