siRNA Details
| Gene Symbol: Coa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195488.1 | NP_001182417.1 | 1 | 150 | ||
| GenBank mRNA | CB607904.1 | - | 1 | 167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coa5| Gene Symbol: Coa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195488.1 | NP_001182417.1 | 1 | 150 | ||
| GenBank mRNA | CB607904.1 | - | 1 | 167 | ||