siRNA Details
| Gene Symbol: Asb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037367.1 | NP_001032444.1 | 3 | 926 | ||
| XM_006257044.2 | XP_006257106.1 | 2 | 879 | |||
| GenBank mRNA | BC107911.1 | - | 3 | 926 | 7453889 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb12| Gene Symbol: Asb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037367.1 | NP_001032444.1 | 3 | 926 | ||
| XM_006257044.2 | XP_006257106.1 | 2 | 879 | |||
| GenBank mRNA | BC107911.1 | - | 3 | 926 | 7453889 | |