siRNA Details
| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012621.1 | NP_001012639.1 | 2 | 260 | ||
| GenBank mRNA | AY911718.1 | - | 2 | 260 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc13a4| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012621.1 | NP_001012639.1 | 2 | 260 | ||
| GenBank mRNA | AY911718.1 | - | 2 | 260 | ||