siRNA Details
| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012621.1 | NP_001012639.1 | 9 | 1009 | ||
| XM_006236283.2 | XP_006236345.1 | 8 | 1529 | |||
| XM_006236284.2 | XP_006236346.1 | 7 | 807 | |||
| GenBank mRNA | AY911718.1 | - | 9 | 1009 | ||
| FQ212433.1 | - | 8 | 1515 | |||
| FQ229668.1 | - | 6 | 550 | |||

Slc13a4
Genomic Map