siRNA Details
| Gene Symbol: Grem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019282.3 | NP_062155.1 | 2 | 480 | ||
| GenBank mRNA | Y10019.1 | - | 2 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grem1| Gene Symbol: Grem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019282.3 | NP_062155.1 | 2 | 480 | ||
| GenBank mRNA | Y10019.1 | - | 2 | 479 | ||