siRNA Details
| Gene Symbol: Nbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031609.1 | NP_113797.1 | 3 | 408 | ||
| XM_008764284.1 | XP_008762506.1 | 3 | 372 | |||
| GenBank mRNA | BC061833.1 | - | 3 | 355 | 5599381 | |
| S72637.1 | - | 3 | 408 | |||
| X66872.1 | - | 3 | 408 | |||

Nbl1
Genomic Map