siRNA Details
| Gene Symbol: Slc6a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017335.1 | NP_059031.1 | 4 | 732 | ||
| GenBank mRNA | FQ229573.1 | - | 256 | |||
| U28927.1 | - | 4 | 732 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a12| Gene Symbol: Slc6a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017335.1 | NP_059031.1 | 4 | 732 | ||
| GenBank mRNA | FQ229573.1 | - | 256 | |||
| U28927.1 | - | 4 | 732 | |||