siRNA Details
| Gene Symbol: Ptpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017336.1 | NP_059032.1 | 19 | 2859 | ||
| XM_006237571.2 | XP_006237633.1 | 8 | 767 | |||
| XM_006237572.2 | XP_006237634.1 | 7 | 681 | |||
| GenBank mRNA | D45412.1 | - | 10 | 1098 | ||
| U28938.1 | - | 19 | 2859 | |||

Ptpro
Genomic Map