siRNA Details
| Gene Symbol: Myl12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017343.2 | NP_059039.2 | 3, 4 | 451 | ||
| GenBank mRNA | BC060577.1 | - | 3, 4 | 450 | 6889406 | |
| EV769828.1 | - | 3, 4 | 438 | |||
| FQ215510.1 | - | 3, 4 | 437 | |||
| FQ229195.1 | - | 3, 4 | 437 | |||
| X52840.1 | - | 2, 3 | 368 | |||

Myl12b
Genomic Map