siRNA Details
| Gene Symbol: Mapk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017347.2 | NP_059043.1 | 3 | 446 | ||
| GenBank mRNA | AF155236.1 | - | 3 | 422 | ||
| M38194.1 | - | 3 | 383 | |||
| M61177.1 | - | 3 | 404 | |||
| S46779.1 | - | 3 | 446 | |||
| U12008.1 | - | 3 | 383 | |||
| X65198.1 | - | 3 | 446 | |||

Mapk3
Genomic Map