siRNA Details
| Gene Symbol: Hpcal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017357.1 | NP_059053.1 | 4 | 589 | ||
| XM_006238829.2 | XP_006238891.1 | 5 | 884 | |||
| GenBank mRNA | D13125.1 | - | 4 | 589 | ||
| FQ212482.1 | - | 5 | 717 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpcal4| Gene Symbol: Hpcal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017357.1 | NP_059053.1 | 4 | 589 | ||
| XM_006238829.2 | XP_006238891.1 | 5 | 884 | |||
| GenBank mRNA | D13125.1 | - | 4 | 589 | ||
| FQ212482.1 | - | 5 | 717 | |||