siRNA Details
| Gene Symbol: Rab10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017359.2 | NP_059055.2 | 2 | 210 | ||
| GenBank mRNA | AY724493.1 | - | 2 | 389 | ||
| BC085744.1 | - | 2 | 210 | 7191921 | ||
| M83677.1 | - | 2 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab10| Gene Symbol: Rab10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017359.2 | NP_059055.2 | 2 | 210 | ||
| GenBank mRNA | AY724493.1 | - | 2 | 389 | ||
| BC085744.1 | - | 2 | 210 | 7191921 | ||
| M83677.1 | - | 2 | 362 | |||