siRNA Details
| Gene Symbol: Rab10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017359.2 | NP_059055.2 | 4, 5 | 470 | ||
| GenBank mRNA | AY310140.1 | - | 2, 3 | 234 | ||
| AY724493.1 | - | 4, 5 | 649 | |||
| BC085744.1 | - | 4, 5 | 470 | 7191921 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab10| Gene Symbol: Rab10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017359.2 | NP_059055.2 | 4, 5 | 470 | ||
| GenBank mRNA | AY310140.1 | - | 2, 3 | 234 | ||
| AY724493.1 | - | 4, 5 | 649 | |||
| BC085744.1 | - | 4, 5 | 470 | 7191921 | ||