siRNA Details
Gene Symbol: Dbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001053826.4 | XP_001053826.1 | 4 | 933 | ||
XM_006242286.2 | XP_006242348.1 | 3 | 837 | |||
GenBank mRNA | AB121147.1 | - | 2 | 239 |
