siRNA Details
| Gene Symbol: Btg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019290.1 | NP_062163.1 | 5 | 691 | ||
| GenBank mRNA | AF087037.1 | - | 5 | 691 | ||
| BC127498.1 | - | 5 | 784 | 8371953 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btg3| Gene Symbol: Btg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019290.1 | NP_062163.1 | 5 | 691 | ||
| GenBank mRNA | AF087037.1 | - | 5 | 691 | ||
| BC127498.1 | - | 5 | 784 | 8371953 | ||