siRNA Details
| Gene Symbol: Cpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019300.1 | NP_062173.1 | 4, 5 | 377 | ||
| GenBank mRNA | FM101110.1 | - | 4, 5 | 377 | ||
| U67914.1 | - | 4, 5 | 363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpa3| Gene Symbol: Cpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019300.1 | NP_062173.1 | 4, 5 | 377 | ||
| GenBank mRNA | FM101110.1 | - | 4, 5 | 377 | ||
| U67914.1 | - | 4, 5 | 363 | |||