siRNA Details
| Gene Symbol: Cr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005265.1 | NP_001005265.1 | 5 | 941 | ||
| NM_001005330.1 | NP_001005330.1 | 941 | ||||
| NM_019301.2 | NP_062174.1 | 5 | 941 | |||
| GenBank mRNA | BC061736.1 | - | 5 | 710 | 5599318 | |
| D42114.1 | - | 5 | 750 | |||
| D42115.1 | - | 5 | 686 | |||
| D42116.1 | - | 2 | 294 | |||
| L36532.1 | - | 5 | 709 | |||

Cr1l
Genomic Map