siRNA Details
| Gene Symbol: Mapk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019319.1 | NP_062192.1 | 1 | 276 | ||
| XM_008772166.1 | XP_008770388.1 | 2 | 1225 | |||
| XM_008772167.1 | XP_008770389.1 | 2 | 1174 | |||
| GenBank mRNA | Z21935.1 | - | 1 | 198 | ||

Mapk4
Genomic Map