siRNA Details
| Gene Symbol: Mcpt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019321.2 | NP_062194.1 | 4 | 496 | ||
| GenBank mRNA | U67907.1 | - | 4 | 498 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcpt4| Gene Symbol: Mcpt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019321.2 | NP_062194.1 | 4 | 496 | ||
| GenBank mRNA | U67907.1 | - | 4 | 498 | ||