siRNA Details
| Gene Symbol: Furin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019331.1 | NP_062204.1 | 6 | 962 | ||
| XM_008759555.1 | XP_008757777.1 | 6 | 846 | |||
| XM_008759556.1 | XP_008757778.1 | 6 | 893 | |||
| GenBank mRNA | X55660.1 | - | 6 | 962 | ||

Furin
Genomic Map