siRNA Details
| Gene Symbol: Prkg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105731.3 | NP_001099201.3 | 14, 15 | 1795 | ||
| GenBank mRNA | EU251189.1 | - | 14, 15 | 1795 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkg1| Gene Symbol: Prkg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105731.3 | NP_001099201.3 | 14, 15 | 1795 | ||
| GenBank mRNA | EU251189.1 | - | 14, 15 | 1795 | ||