siRNA Details
| Gene Symbol: Ezr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019357.1 | NP_062230.1 | 6 | 1151 | ||
| GenBank mRNA | AF450298.1 | - | 6 | 750 | ||
| AY428869.1 | - | 6 | 736 | |||
| BC081958.1 | - | 6 | 1151 | 7124292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ezr| Gene Symbol: Ezr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019357.1 | NP_062230.1 | 6 | 1151 | ||
| GenBank mRNA | AF450298.1 | - | 6 | 750 | ||
| AY428869.1 | - | 6 | 736 | |||
| BC081958.1 | - | 6 | 1151 | 7124292 | ||