siRNA Details
Gene Symbol: Crlf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168612.1 | NP_001162083.1 | 1 | 184 | ||
GenBank mRNA | CB611803.1 | - | 184 | |||
CO555844.1 | - | 1 | 183 | |||
DY321186.1 | - | 1 | 177 |
