siRNA Details
| Gene Symbol: Ywhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019376.2 | NP_062249.1 | 1 | 157 | ||
| XM_006249184.2 | XP_006249246.1 | 1 | 285 | |||
| GenBank mRNA | BC127496.1 | - | 1 | 157 | 8372306 | |
| D17447.1 | - | 1 | 120 | |||
| S55305.1 | - | 1 | 120 | |||

Ywhag
Genomic Map