siRNA Details
| Gene Symbol: Gorasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019385.2 | NP_062258.2 | 5 | 519 | ||
| GenBank mRNA | AF015264.2 | - | 5 | 496 | ||
| BC081809.1 | - | 5 | 513 | |||
| BC161850.1 | - | 5 | 519 | 8369057 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gorasp1| Gene Symbol: Gorasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019385.2 | NP_062258.2 | 5 | 519 | ||
| GenBank mRNA | AF015264.2 | - | 5 | 496 | ||
| BC081809.1 | - | 5 | 513 | |||
| BC161850.1 | - | 5 | 519 | 8369057 | ||