siRNA Details
| Gene Symbol: Mrpl37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004235.1 | NP_001004235.1 | 6 | 1142 | ||
| GenBank mRNA | BC079328.1 | - | 6 | 1142 | 7191681 | |
| FQ215540.1 | - | 6 | 1155 | |||
| FQ226708.1 | - | 4 | 522 | |||
| FQ231717.1 | - | 4 | 522 | |||

Mrpl37
Genomic Map