siRNA Details
Gene Symbol: LOC56764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020089.2 | NP_064474.1 | 1 | 460 | ||
GenBank mRNA | AF154849.1 | - | 1 | 434 | ||
BC083558.1 | - | 1 | 460 | 7134067 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC56764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020089.2 | NP_064474.1 | 1 | 460 | ||
GenBank mRNA | AF154849.1 | - | 1 | 434 | ||
BC083558.1 | - | 1 | 460 | 7134067 |