siRNA Details
| Gene Symbol: Ly6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020103.1 | NP_064488.1 | 1, 2 | 80 | ||
| GenBank mRNA | M30691.1 | - | 1, 2 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6c| Gene Symbol: Ly6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020103.1 | NP_064488.1 | 1, 2 | 80 | ||
| GenBank mRNA | M30691.1 | - | 1, 2 | 78 | ||