siRNA Details
| Gene Symbol: Ramp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020100.2 | NP_064485.2 | 3 | 307 | ||
| GenBank mRNA | AB028935.1 | - | 2 | 108 | ||
| AB030944.1 | - | 3 | 286 | |||
| AB042889.1 | - | 3 | 305 | |||
| AF181552.2 | - | 3 | 307 | |||
| FQ214551.1 | - | 3 | 301 | |||
| FQ220330.1 | - | 3 | 288 | |||
| FQ220789.1 | - | 3 | 309 | |||
| FQ229563.1 | - | 3 | 326 | |||

Ramp3
Genomic Map