siRNA Details
| Gene Symbol: Ramp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020100.2 | NP_064485.2 | 1, 2 | 90 | ||
| GenBank mRNA | AB030944.1 | - | 1, 2 | 69 | ||
| AB042889.1 | - | 1, 2 | 88 | |||
| AF181552.2 | - | 1, 2 | 90 | |||
| FQ214551.1 | - | 1, 2 | 84 | |||
| FQ220330.1 | - | 1, 2 | 71 | |||
| FQ220789.1 | - | 1, 2 | 92 | |||
| FQ229563.1 | - | 1, 2 | 109 | |||

Ramp3
Genomic Map