siRNA Details
| Gene Symbol: Adam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020305.1 | NP_064701.1 | 1 | 2216 | ||
| GenBank mRNA | BC107437.1 | - | 1 | 2216 | 7132937 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam4| Gene Symbol: Adam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020305.1 | NP_064701.1 | 1 | 2216 | ||
| GenBank mRNA | BC107437.1 | - | 1 | 2216 | 7132937 | |