siRNA Details
| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020656.2 | NP_065707.2 | 9 | 749 | ||
| GenBank mRNA | AF264765.2 | - | 9 | 749 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parva| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020656.2 | NP_065707.2 | 9 | 749 | ||
| GenBank mRNA | AF264765.2 | - | 9 | 749 | ||