siRNA Details
| Gene Symbol: Nckap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031618.1 | NP_113806.1 | 2261 | |||
| GenBank mRNA | BC092610.1 | - | 3 | 214 | ||
| BC107935.1 | - | 8 | 821 | |||
| D84346.1 | - | 20 | 2165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nckap1| Gene Symbol: Nckap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031618.1 | NP_113806.1 | 2261 | |||
| GenBank mRNA | BC092610.1 | - | 3 | 214 | ||
| BC107935.1 | - | 8 | 821 | |||
| D84346.1 | - | 20 | 2165 | |||