siRNA Details
| Gene Symbol: Prg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031619.2 | NP_113807.2 | 2, 3 | 111 | ||
| GenBank mRNA | CB703061.1 | - | 1, 2 | 106 | ||
| D50568.1 | - | 2, 3 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prg2| Gene Symbol: Prg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031619.2 | NP_113807.2 | 2, 3 | 111 | ||
| GenBank mRNA | CB703061.1 | - | 1, 2 | 106 | ||
| D50568.1 | - | 2, 3 | 111 | |||