siRNA Details
Gene Symbol: Prg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031619.2 | NP_113807.2 | 5 | 587 | ||
GenBank mRNA | CA509460.1 | - | 2 | 268 | ||
D50568.1 | - | 5 | 587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031619.2 | NP_113807.2 | 5 | 587 | ||
GenBank mRNA | CA509460.1 | - | 2 | 268 | ||
D50568.1 | - | 5 | 587 |