siRNA Details
| Gene Symbol: Robo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022188.1 | NP_071524.1 | 5 | 762 | ||
| GenBank mRNA | AF041082.1 | - | 5 | 762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Robo1| Gene Symbol: Robo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022188.1 | NP_071524.1 | 5 | 762 | ||
| GenBank mRNA | AF041082.1 | - | 5 | 762 | ||