siRNA Details
Gene Symbol: Gabarap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172036.3 | NP_742033.1 | 3 | 266 | ||
GenBank mRNA | AF161588.1 | - | 3 | 280 | ||
BC058441.1 | - | 3 | 270 | 6918795 | ||
FQ224284.1 | - | 3 | 232 | |||
FQ229256.1 | - | 3 | 268 |
