siRNA Details
| Gene Symbol: Gabarap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172036.3 | NP_742033.1 | 1 | 74 | ||
| GenBank mRNA | AF161588.1 | - | 1 | 88 | ||
| BC058441.1 | - | 1 | 78 | 6918795 | ||
| FQ213199.1 | - | 1 | 145 | |||
| FQ224284.1 | - | 1 | 40 | |||
| FQ229256.1 | - | 1 | 76 | |||

Gabarap
Genomic Map