siRNA Details
| Gene Symbol: Rabggta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031654.2 | NP_113842.1 | 12 | 1183 | ||
| XM_006252009.2 | XP_006252071.1 | 12 | 1278 | |||
| XM_006252010.2 | XP_006252072.1 | 12 | 1225 | |||
| GenBank mRNA | BC086547.1 | - | 12 | 1183 | 7301723 | |
| L10415.1 | - | 1143 | ||||
| S62096.1 | - | 1143 | ||||

Rabggta
Genomic Map