siRNA Details
| Gene Symbol: Prl8a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021580.1 | NP_067591.1 | 4 | 357 | ||
| XM_008771599.1 | XP_008769821.1 | 4 | 293 | |||
| GenBank mRNA | AB009889.1 | - | 4 | 357 | ||
| Gene Symbol: LOC103694085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771616.1 | XP_008769838.1 | 299 | |||
| XM_008771617.1 | XP_008769839.1 | 293 | ||||

Prl8a4
Genomic Map