siRNA Details
| Gene Symbol: Ltbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021587.1 | NP_067598.1 | 23 | 3833 | ||
| GenBank mRNA | M55431.1 | - | 23 | 3833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltbp1| Gene Symbol: Ltbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021587.1 | NP_067598.1 | 23 | 3833 | ||
| GenBank mRNA | M55431.1 | - | 23 | 3833 | ||