siRNA Details
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021589.1 | NP_067600.1 | 14 | 1762 | ||
| XM_006232746.2 | XP_006232808.1 | 13 | 1735 | |||
| GenBank mRNA | M85214.1 | - | 14 | 1762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntrk1| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021589.1 | NP_067600.1 | 14 | 1762 | ||
| XM_006232746.2 | XP_006232808.1 | 13 | 1735 | |||
| GenBank mRNA | M85214.1 | - | 14 | 1762 | ||