siRNA Details
| Gene Symbol: Slc9a3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021594.1 | NP_067605.1 | 5 | 883 | ||
| GenBank mRNA | AF154336.1 | - | 5 | 883 | ||
| FQ213792.1 | - | 5 | 1000 | |||
| FQ228926.1 | - | 5 | 981 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a3r1| Gene Symbol: Slc9a3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021594.1 | NP_067605.1 | 5 | 883 | ||
| GenBank mRNA | AF154336.1 | - | 5 | 883 | ||
| FQ213792.1 | - | 5 | 1000 | |||
| FQ228926.1 | - | 5 | 981 | |||