siRNA Details
| Gene Symbol: Ghrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021669.2 | NP_067701.1 | 3, 4 | 362 | ||
| GenBank mRNA | AB029433.1 | - | 3, 4 | 363 | ||
| AB035699.1 | - | 3, 4 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ghrl| Gene Symbol: Ghrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021669.2 | NP_067701.1 | 3, 4 | 362 | ||
| GenBank mRNA | AB029433.1 | - | 3, 4 | 363 | ||
| AB035699.1 | - | 3, 4 | 356 | |||