siRNA Details
| Gene Symbol: Tmem33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034198.2 | NP_001029370.1 | 7, 8 | 773 | ||
| NM_021671.3 | NP_067703.1 | 7, 8 | 773 | |||
| GenBank mRNA | AB006135.1 | - | 7, 8 | 731 | ||
| BF552634.1 | - | 2, 3 | 105 | |||
| CK359233.1 | - | 7, 8 | 654 | |||

Tmem33
Genomic Map