siRNA Details
| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021676.1 | NP_067708.1 | 21 | 3802 | ||
| GenBank mRNA | AF133301.1 | - | 21 | 3802 | ||
| AJ133120.1 | - | 21 | 3803 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shank3| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021676.1 | NP_067708.1 | 21 | 3802 | ||
| GenBank mRNA | AF133301.1 | - | 21 | 3802 | ||
| AJ133120.1 | - | 21 | 3803 | |||