siRNA Details
Gene Symbol: Shank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021676.1 | NP_067708.1 | 21 | 4783 | ||
GenBank mRNA | AF133301.1 | - | 21 | 4783 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Shank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021676.1 | NP_067708.1 | 21 | 4783 | ||
GenBank mRNA | AF133301.1 | - | 21 | 4783 |