siRNA Details
| Gene Symbol: Nxph4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021680.2 | NP_067712.2 | 1 | 61 | ||
| GenBank mRNA | AF042714.1 | - | 1 | 132 | ||
| BC081805.1 | - | 1 | 61 | 7108207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxph4| Gene Symbol: Nxph4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021680.2 | NP_067712.2 | 1 | 61 | ||
| GenBank mRNA | AF042714.1 | - | 1 | 132 | ||
| BC081805.1 | - | 1 | 61 | 7108207 | ||